missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,3-thiadiazole-5-carboxylic acid, 97%, Thermo Scientific™

11.30 € - 113.00 €

Chemical Identifiers

CAS 18212-21-0
Molecular Formula C4H3N2O2S
Molecular Weight (g/mol) 143.14
MDL Number MFCD00173830
InChI Key NHHQOYLPBUYHQU-UHFFFAOYSA-M
Synonym 4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
PubChem CID 1381992
IUPAC Name 4-methylthiadiazole-5-carboxylic acid
SMILES CC1=C(SN=N1)C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10079091
View Documents
Thermo Scientific Maybridge
CC02301DA
1 g
11.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
10476604
View Documents
Thermo Scientific Maybridge
CC02301EA
10 g
51.50 €
10g
Please to purchase this item. Need a web account? Register with us today!
10396974
View Documents
Thermo Scientific Maybridge
CC02301EB
25 g
113.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

18212-21-0
143.14
NHHQOYLPBUYHQU-UHFFFAOYSA-M
1381992
CC1=C(SN=N1)C([O-])=O
C4H3N2O2S
MFCD00173830
4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
4-methylthiadiazole-5-carboxylic acid

Specifications

18212-21-0
100.0
Amber glass bottle
MFCD00173830
NHHQOYLPBUYHQU-UHFFFAOYSA-M
4-methylthiadiazole-5-carboxylic acid
1381992
97%
96.5
97%
C4H3N2O2S
4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
CC1=C(SN=N1)C([O-])=O
143.14
144.15
4-Methyl-1, 2, 3-thiadiazole-5-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.