Learn More
4-Methyl-1,2-cyclohexene oxide, cis + trans, 97%, Thermo Scientific Chemicals
33.70 € - 334.00 €
Chemical Identifiers
CAS | 36099-51-1 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.172 |
MDL Number | MFCD09742280 |
InChI Key | ULPDSNLBZMHGPI-UHFFFAOYSA-N |
Synonym | 3-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane, 3-methyl, 4-methyl-1,2-cyclohexene oxide, 4-methyl-1,2-cyclohexene oxide, cis + trans, 4-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane,3-methyl, 3-methyl-7-oxabicyclo 4.1.0 heptane #, 4-methyl-1,2-cyclohexene oxide, cis+trans |
PubChem CID | 535184 |
IUPAC Name | 4-methyl-7-oxabicyclo[4.1.0]heptane |
SMILES | CC1CCC2C(C1)O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11422230
|
Thermo Scientific Alfa Aesar
H29100.03 |
1 g |
33.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432230
|
Thermo Scientific Alfa Aesar
H29100.06 |
5 g |
104.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442230
|
Thermo Scientific Alfa Aesar
H29100.14 |
25 g |
334.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
36099-51-1 | |
112.172 | |
ULPDSNLBZMHGPI-UHFFFAOYSA-N | |
535184 | |
CC1CCC2C(C1)O2 |
C7H12O | |
MFCD09742280 | |
3-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane, 3-methyl, 4-methyl-1,2-cyclohexene oxide, 4-methyl-1,2-cyclohexene oxide, cis + trans, 4-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane,3-methyl, 3-methyl-7-oxabicyclo 4.1.0 heptane #, 4-methyl-1,2-cyclohexene oxide, cis+trans | |
4-methyl-7-oxabicyclo[4.1.0]heptane |
Specifications
36099-51-1 | |
1.447 | |
3-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane, 3-methyl, 4-methyl-1,2-cyclohexene oxide, 4-methyl-1,2-cyclohexene oxide, cis + trans, 4-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane,3-methyl, 3-methyl-7-oxabicyclo 4.1.0 heptane #, 4-methyl-1,2-cyclohexene oxide, cis+trans | |
CC1CCC2C(C1)O2 | |
112.172 | |
112.17 | |
4-Methyl-1,2-cyclohexene oxide, Cis + Trans |
C7H12O | |
MFCD09742280 | |
ULPDSNLBZMHGPI-UHFFFAOYSA-N | |
4-methyl-7-oxabicyclo[4.1.0]heptane | |
535184 | |
97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 4-Methyl-1,2-cyclohexene oxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 4-Methyl-1,2-cyclohexene oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.