missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-oxazol-2-amine, Thermo Scientific™

Product Code. 10099401
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35629-70-0
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00126682
InChI Key VCZJVXLWQTXSPQ-UHFFFAOYSA-N
PubChem CID 535824
IUPAC Name 4-methyl-1,3-oxazol-2-amine
SMILES CC1=COC(=N1)N

Specifications

Packaging Amber glass bottle
Quantity 1 g
Synonym 4-methyloxazol-2-amine, 2-amino-4-methyloxazole, 2-oxazolamine, 4-methyl, 4-methyl-oxazol-2-ylamine, 2-oxazolamine,4-methyl, 2-amino-4-methyl-1,3-oxazole, 2-amino-4-methyloxazol, 4-methyl-2-oxazolamine, 4-methyl-2-amino-oxazole, 2-amino-4-methyl-oxazole
Formula Weight 98.1
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-Methyl-1, 3-oxazol-2-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'