Learn More
4-Methyl-1-piperazinepropionitrile, 98%, Thermo Scientific™
47.70 € - 191.96 €
Chemical Identifiers
CAS | 4491-92-3 |
---|---|
Molecular Formula | C8H15N3 |
Molecular Weight (g/mol) | 153.229 |
MDL Number | MFCD00051960 |
InChI Key | XMZMOGFGIJQCHN-UHFFFAOYSA-N |
Synonym | 3-4-methylpiperazin-1-yl propanenitrile, 1-2-cyanoethyl-4-methylpiperazine, 3-4-methylpiperazinyl propanenitrile, acmc-1cumv, 4-methyl-1-piperazinepropionitrile, 4-methyl-1-piperazinepropanenitrile, 1-piperazinepropanenitrile, 4-methyl, n-2-cyanoethyl-n'-methylpiperazine, 3-4-methyl-piperazin-1-yl-propionitrile |
PubChem CID | 3050146 |
IUPAC Name | 3-(4-methylpiperazin-1-yl)propanenitrile |
SMILES | CN1CCN(CC1)CCC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382776
|
Thermo Scientific Alfa Aesar
L08905.09 |
10 g | N/A | ||||||
11392776
|
Thermo Scientific Alfa Aesar
L08905.18 |
50 g |
191.96 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
4491-92-3 | |
153.229 | |
XMZMOGFGIJQCHN-UHFFFAOYSA-N | |
3050146 | |
CN1CCN(CC1)CCC#N |
C8H15N3 | |
MFCD00051960 | |
3-4-methylpiperazin-1-yl propanenitrile, 1-2-cyanoethyl-4-methylpiperazine, 3-4-methylpiperazinyl propanenitrile, acmc-1cumv, 4-methyl-1-piperazinepropionitrile, 4-methyl-1-piperazinepropanenitrile, 1-piperazinepropanenitrile, 4-methyl, n-2-cyanoethyl-n'-methylpiperazine, 3-4-methyl-piperazin-1-yl-propionitrile | |
3-(4-methylpiperazin-1-yl)propanenitrile |
Specifications
4491-92-3 | |
C8H15N3 | |
MFCD00051960 | |
117453 | |
XMZMOGFGIJQCHN-UHFFFAOYSA-N | |
3-(4-methylpiperazin-1-yl)propanenitrile | |
3050146 | |
98% |
134°C to 136°C (13 mmHg) | |
1.476 | |
UN3276 | |
3-4-methylpiperazin-1-yl propanenitrile, 1-2-cyanoethyl-4-methylpiperazine, 3-4-methylpiperazinyl propanenitrile, acmc-1cumv, 4-methyl-1-piperazinepropionitrile, 4-methyl-1-piperazinepropanenitrile, 1-piperazinepropanenitrile, 4-methyl, n-2-cyanoethyl-n'-methylpiperazine, 3-4-methyl-piperazin-1-yl-propionitrile | |
CN1CCN(CC1)CCC#N | |
153.229 | |
153.23 | |
4-Methyl-1-piperazinepropionitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures