missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10522473
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 144059-86-9
Molecular Formula C12H8F3NO2S
Molecular Weight (g/mol) 287.256
MDL Number MFCD00068105
InChI Key DRFFZMPSUPHSJN-UHFFFAOYSA-N
Synonym 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxylic acid, 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-4-trifluoromethylphenyl thiazole-5-carboxylic acid, 2-4-trifluoromethyl phenyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-4-trifluoromethyl-phenyl-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 4-methyl-2-4-trifluoromethyl phenyl, 5-thiazolecarboxylicacid, 4-methyl-2-4-trifluoromethyl phenyl, maybridge1_008801
PubChem CID 2775663
IUPAC Name 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 287.26
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.