missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitrophenol, 97%, Thermo Scientific Chemicals

44.50 € - 545.00 €

Chemical Identifiers

CAS 119-33-5
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007120
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
PubChem CID 8391
IUPAC Name 4-methyl-2-nitrophenol
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459852
View Documents
Thermo Scientific Alfa Aesar
A12009.14
25 g
44.50 €
25g
Please to purchase this item. Need a web account? Register with us today!
11469852
View Documents
Thermo Scientific Alfa Aesar
A12009.22
100 g
138.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
11479852
View Documents
Thermo Scientific Alfa Aesar
A12009.36
500 g
545.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119-33-5
153.137
SYDNSSSQVSOXTN-UHFFFAOYSA-N
8391
CC1=CC(=C(C=C1)O)[N+](=O)[O-]
C7H7NO3
MFCD00007120
2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
4-methyl-2-nitrophenol

Specifications

119-33-5
1.24
108°C (226°F)
C7H7NO3
1868022
SYDNSSSQVSOXTN-UHFFFAOYSA-N
4-methyl-2-nitrophenol
8391
97%
32°C to 35°C
125°C (22 mmHg)
Phenol-like
MFCD00007120
2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
CC1=CC(=C(C=C1)O)[N+](=O)[O-]
153.137
153.14
4-Methyl-2-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 204-315-6

missing translation for 'rtecsNumber' : GP2800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only