missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, CHROMASOLV™, for HPLC, ≥99.5%, Honeywell Riedel-de Haen

Product Code. 15652960
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

CHROMASOLV™, for HPLC,≥99.5%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-10-1
Absorbance max. 0.01 at 400 nm, max. 0.02 at 380 nm, max. 0.15 at 360 nm, max. 0.50 at 340 nm, max. 1.0 at 335 nm
CAS Max % 100.0000%
Melting Point -80°C (literature)
Density 0.801 g/mL at 25 °C(lit.)
Boiling Point 115°C to 117°C
Flash Point 14°C
Packaging Glass Bottle
Molecular Formula C6H12O
Linear Formula (CH3)2CHCH2COCH3
MDL Number MFCD00008938
Quantity 1 L
UN Number UN1245
Beilstein 605399
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
SMILES CC(C)CC(=O)C
IUPAC Name 4-methylpentan-2-one
Molecular Weight (g/mol) 100.161
PubChem CID 7909
ChEBI CHEBI:82344
Formula Weight 100.16
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material 4-Methyl-2-pentanone, CHROMASOLV™, for HPLC, ≥99.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-2-pentanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.