missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, puriss. p.a., ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
missing translation for 'orderingAttributeHoverText'
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Förpackningsstorlek
Pack of 4
Pack of 6
Beskrivelse
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Tekniske data
Tekniske data
| Melting Point | -80°C |
| Color | Colorless |
| Density | 0.801g/mL at 25°C |
| Boiling Point | 117°C |
| Flash Point | 14°C (57.2°F) |
| Odor | Characteristic |
| Packaging | Glass Bottle |
| Linear Formula | (CH3)2CHCH2COCH3 |
| Quantity | 6 x 1 L |
| UN Number | UN1245 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion