missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-thiazolecarboxaldehyde, 97%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Boiling Point | 90.0°C to 92.0°C (12.0 mbar) |
Flash Point | 67°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Quantity | 1 g |
Formula Weight | 127.17 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methyl-2-thiazolecarboxaldehyde |
Sicherheit und Handhabung
Produktidentifikator
- 4-Methyl-2-thiazolecarboxaldehyde
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'