missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine, 90%, Thermo Scientific™

131.60 € - 163.80 €

Chemical Identifiers

CAS 915707-59-4
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD09065018
InChI Key OGPGKFFHFGZOBY-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methylamine, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazin-7-yl methanamine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 2h-pyrido 3,2-b-1,4-oxazine-7-methanamine,3,4-dihydro-4-methyl
PubChem CID 24229650
IUPAC Name (4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanamine
SMILES CN1CCOC2=C1N=CC(=C2)CN
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10143174
View Documents Promotion Details
Thermo Scientific Maybridge
CC62013CB
250 mg Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10474813
View Documents Promotion Details
Thermo Scientific Maybridge
CC62013DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

915707-59-4
179.223
OGPGKFFHFGZOBY-UHFFFAOYSA-N
24229650
CN1CCOC2=C1N=CC(=C2)CN
C9H13N3O
MFCD09065018
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methylamine, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazin-7-yl methanamine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 2h-pyrido 3,2-b-1,4-oxazine-7-methanamine,3,4-dihydro-4-methyl
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanamine

Specifications

915707-59-4
100.0
C9H13N3O
4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methylamine, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazin-7-yl methanamine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 2h-pyrido 3,2-b-1,4-oxazine-7-methanamine,3,4-dihydro-4-methyl
CN1CCOC2=C1N=CC(=C2)CN
179.223
179.22
(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine
89.5
90%
MFCD09065018
OGPGKFFHFGZOBY-UHFFFAOYSA-N
(4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanamine
24229650
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications