missing translation for 'onlineSavingsMsg'
Learn More

4-Methylaminophenol sulfate, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10479070
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-55-0
Molecular Formula C14H18N2O2·H2SO4
Molecular Weight (g/mol) 344.38
MDL Number MFCD00013140
Synonym Metol

Specifications

CAS Min % 98.5
CAS Max % 100.0
Decomposition Information Decomposition Information: >260°C
Melting Point 260.0°C
Color Beige to White or Gray
Density 1.5770g/mL
Ignition Residue 0.15% max.
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3NHC6H4OH)2·H2SO4
Quantity 5 g
Beilstein 13, 441
Merck Index 15, 6091
Specific Gravity 1.577
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in 20 parts cold water,soluble in 6 parts boiling water,slightly soluble in alcohol,insoluble in ether
Formula Weight 344.38
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylaminophenol sulfate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methylaminophenol sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.