missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbenzoylacetonitrile, 99%, Thermo Scientific™
Click to view available options
1g; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
CAS | 7391-28-8 |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 104.0°C to 108.0°C |
Color | Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C10H9NO |
MDL Number | MFCD00067895 |
Show More |
Safety and Handling
Product Identifier
- 4-Methylbenzoylacetonitrile
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction