missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanol, cis + trans, 98%, Thermo Scientific Chemicals

Product Code. 11482738
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-91-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001449,MFCD00064171,MFCD00064170
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(O)CC1

Specifications

Melting Point -41°C
Density 0.915
Boiling Point 171°C to 173°C
Flash Point 70°C (158°F)
Refractive Index 1.458
Quantity 1000 mL
Beilstein 1445998
Solubility Information Soluble in water (15mg/ml at 20°C),ether,alcohols,and many organic solvents.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 4-Methylcyclohexanol, cis + trans
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'