Learn More
4-Methylcyclohexanol, cis + trans, 98%, Thermo Scientific Chemicals
26.60 € - 165.00 €
Chemical Identifiers
CAS | 589-91-3 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.19 |
MDL Number | MFCD00001449,MFCD00064171,MFCD00064170 |
InChI Key | MQWCXKGKQLNYQG-UHFFFAOYSA-N |
Synonym | 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol |
PubChem CID | 11524 |
IUPAC Name | 4-methylcyclohexan-1-ol |
SMILES | CC1CCC(O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11492738
|
Thermo Scientific Alfa Aesar
A16734.AD |
50 mL |
26.60 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402748
|
Thermo Scientific Alfa Aesar
A16734.AK |
250 mL |
53.20 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11482738
|
Thermo Scientific Alfa Aesar
A16734.AU |
1000 mL |
165.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.
Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.
Chemical Identifiers
589-91-3 | |
114.19 | |
MQWCXKGKQLNYQG-UHFFFAOYSA-N | |
11524 | |
CC1CCC(O)CC1 |
C7H14O | |
MFCD00001449,MFCD00064171,MFCD00064170 | |
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol | |
4-methylcyclohexan-1-ol |
Specifications
589-91-3 | |
0.915 | |
70°C (158°F) | |
1.458 | |
1445998 | |
Soluble in water (15mg/ml at 20°C),ether,alcohols,and many organic solvents. | |
CC1CCC(O)CC1 | |
114.19 | |
114.19 | |
4-Methylcyclohexanol cis + trans, cis + trans |
°C to 41°C | |
171°C to 173°C | |
C7H14O | |
MFCD00001449,MFCD00064171,MFCD00064170 | |
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol | |
MQWCXKGKQLNYQG-UHFFFAOYSA-N | |
4-methylcyclohexan-1-ol | |
11524 | |
98% |
Safety and Handling
GHS H Statement
H227
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c
H227-H302+H312+H332-H315-H320-H335
missing translation for 'einecsNumber' : 209-664-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only