missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Product Code. 11440753
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 940-64-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00014365
InChI Key SFTDDFBJWUWKMN-UHFFFAOYSA-N
Synonym 4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
PubChem CID 70329
IUPAC Name 2-(4-methylphenoxy)acetic acid
SMILES CC1=CC=C(C=C1)OCC(=O)O

Specifications

Melting Point 137°C to 142°C
Quantity 10 g
Beilstein 2047591
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 4-Methylphenoxyacetic acid
compliance-icons
Product Identifier
  • (4-Methylphenoxy)acetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : (4-Methylphenoxy)acetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'