missing translation for 'onlineSavingsMsg'
Learn More
4-Methylpiperidine, 98+%, Thermo Scientific Chemicals Product Code.: 11317805

4-Methylpiperidine, 98+%, Thermo Scientific Chemicals

Product Code. 11317805
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11317805

Brand: Thermo Scientific Alfa Aesar L02709.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-58-4
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00006005
InChI Key UZOFELREXGAFOI-UHFFFAOYSA-N
Synonym 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline
PubChem CID 69381
IUPAC Name 4-methylpiperidine
SMILES CC1CCNCC1

Specifications

Melting Point 4°C to 5°C
Density 0.838
pH 13
Boiling Point 123°C to 124°C
Flash Point 13°C (55°F)
Odor Amine-like
Refractive Index 1.4458
Quantity 500 mL
UN Number UN2733
Beilstein 1083
Solubility Information Miscible with water and chloroform.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material 4-Methylpiperidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methylpiperidine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.