missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfinyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11354805
100 mg
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166386-48-7
Molecular Formula C7H9BO3S
Molecular Weight (g/mol) 184.02
MDL Number MFCD02093071
InChI Key YOTGALZTDVXUKZ-UHFFFAOYNA-N
Synonym 4-methanesulfinyl benzeneboronic acid, 4-methylsulfinyl phenyl boronic acid, 4-methylsulphinyl benzeneboronic acid, 4-methylsulfinylphenyl boronic acid, 4-methanesulphinylphenylboronic acid, 4-methylsulfinyl phenylboronic acid, boronic acid, 4-methylsulfinyl phenyl, 4-boronophenyl methyl sulfoxide, 4-methanesulfinylphenylboronic acid
PubChem CID 2773531
IUPAC Name (4-methylsulfinylphenyl)boronic acid
SMILES CS(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point ∼170°C
Quantity 100 mg
Formula Weight 184.02
Percent Purity 98%
Chemical Name or Material 4-(Methylsulfinyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Methylsulfinyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.