missing translation for 'onlineSavingsMsg'
Learn More

4-Methyltetrahydro-1(2H)-pyrazinecarboximidamide hydroiodide, 97%, Thermo Scientific™

83.70 € - 298.00 €

Chemical Identifiers

CAS 77723-03-6
Molecular Formula C6H15IN4
Molecular Weight (g/mol) 270.118
MDL Number MFCD09025920
InChI Key KESCAEZAONDSRV-UHFFFAOYSA-N
Synonym 4-methylpiperazine-1-carboximidamide hydroiodide, 4-methyltetrahydro-1 2h-pyrazinecarboximidamide hydroiodide, 4-methylpiperazinecarboxamidine, iodide, 4-methyl-1-piperazinecarboximidamide hydroiodide, 4-methylpiperazine-1-carboximidamide-hydrogen iodide 1/1
PubChem CID 20284359
IUPAC Name 4-methylpiperazine-1-carboximidamide;hydroiodide
SMILES CN1CCN(CC1)C(=N)N.I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10442844
View Documents
Thermo Scientific Maybridge
MO01752DA
1 g
83.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
11386885
View Documents
Thermo Scientific Maybridge
MO01752EA
10 g
298.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

77723-03-6
270.118
KESCAEZAONDSRV-UHFFFAOYSA-N
20284359
CN1CCN(CC1)C(=N)N.I
C6H15IN4
MFCD09025920
4-methylpiperazine-1-carboximidamide hydroiodide, 4-methyltetrahydro-1 2h-pyrazinecarboximidamide hydroiodide, 4-methylpiperazinecarboxamidine, iodide, 4-methyl-1-piperazinecarboximidamide hydroiodide, 4-methylpiperazine-1-carboximidamide-hydrogen iodide 1/1
4-methylpiperazine-1-carboximidamide;hydroiodide

Specifications

77723-03-6
97%
C6H15IN4
4-methylpiperazine-1-carboximidamide hydroiodide, 4-methyltetrahydro-1 2h-pyrazinecarboximidamide hydroiodide, 4-methylpiperazinecarboxamidine, iodide, 4-methyl-1-piperazinecarboximidamide hydroiodide, 4-methylpiperazine-1-carboximidamide-hydrogen iodide 1/1
CN1CCN(CC1)C(=N)N.I
270.118
270.11
4-Methyltetrahydro-1(2H)-pyrazinecarboximidamide hydroiodide
97.0
Amber glass bottle
MFCD09025920
KESCAEZAONDSRV-UHFFFAOYSA-N
4-methylpiperazine-1-carboximidamide;hydroiodide
20284359
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.