missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylthio)-6-pyrazin-2-yl-1,3,5-triazin-2-amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Quantity | 10 g |
| Formula Weight | 220.26 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(Methylthio)-6-pyrazin-2-yl-1, 3, 5-triazin-2-amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction