Learn More
4-Methylthiomorpholine 1,1-dioxide, 97%, Thermo Scientific™
54.00 € - 193.00 €
Chemical Identifiers
CAS | 25343-91-3 |
---|---|
Molecular Formula | C5H11NO2S |
Molecular Weight (g/mol) | 149.208 |
MDL Number | MFCD00014623 |
InChI Key | ULZCOWMSBOJCLT-UHFFFAOYSA-N |
PubChem CID | 91413 |
IUPAC Name | 4-methyl-1,4-thiazinane 1,1-dioxide |
SMILES | CN1CCS(=O)(=O)CC1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesKeep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
25343-91-3 | |
149.208 | |
ULZCOWMSBOJCLT-UHFFFAOYSA-N | |
4-methyl-1,4-thiazinane 1,1-dioxide |
C5H11NO2S | |
MFCD00014623 | |
91413 | |
CN1CCS(=O)(=O)CC1 |
Specifications
25343-91-3 | |
C5H11NO2S | |
114527 | |
ULZCOWMSBOJCLT-UHFFFAOYSA-N | |
4-methyl-1,4-thiazinane 1,1-dioxide | |
91413 | |
97% |
84°C to 88°C | |
MFCD00014623 | |
4-methylthiomorpholine 1,1-dioxide, 4-methylthiomorpholine-1,1-dioxide, thiomorpholine, 4-methyl-, 1,1-dioxide, n-methyltetrahydro-1,4-thiazine-s-oxide, 4-methyl-1??-thiomorpholine-1,1-dione, 4-methylthiomorpholine1,1-dioxide, acmc-1cc0z, wln: t6n dswtj a1, 4-27-00-00636 beilstein handbook reference, thiomorpholine,4-methyl-, 1,1-dioxide | |
CN1CCS(=O)(=O)CC1 | |
149.208 | |
149.21 | |
4-Methylthiomorpholine 1,1-dioxide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 246-877-5
missing translation for 'rtecsNumber' : XM3015000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures