Learn More
4-Methylumbelliferyl sulfate potassium salt, 98%, Thermo Scientific Chemicals
Substrate for sulfatase
111.00 € - 715.00 €
Chemical Identifiers
CAS | 15220-11-8 |
---|---|
Molecular Formula | C10H7KO6S |
Molecular Weight (g/mol) | 294.318 |
MDL Number | MFCD00016970 |
InChI Key | CSOCSPXOODWGLJ-UHFFFAOYSA-M |
Synonym | 4-methylumbelliferyl sulfate potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt, 4-methylumbelliferyl sulfate, potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt 1:1, potassium 4-methyl-2-oxo-2h-chromen-7-yl sulfate, potassium 4-methyl-2-oxochromen-7-yl sulfate, c10h7o6s.k, potassium 4-methyl-2-oxo-2h-1-benzopyran-7-yl sulphate, potassium 4-methylumbelliferyl sulfate |
PubChem CID | 5044226 |
IUPAC Name | potassium;(4-methyl-2-oxochromen-7-yl) sulfate |
SMILES | CC1=CC(=O)OC2=C1C=CC(=C2)OS(=O)(=O)[O-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410517
|
Thermo Scientific Alfa Aesar
J66160.ME |
500 mg |
111.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487409
|
Thermo Scientific Alfa Aesar
J66160.03 |
1 g |
185.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15497409
|
Thermo Scientific Alfa Aesar
J66160.06 |
5 g | N/A | ||||||
Description
4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methylumbelliferyl sulfate potassium salt is a fluorometric substrate for the determination of sulfatase. This substrate has been used in the discovery of tyrosine sulfotransferase inhibitors, particularly in P-selectin glycoprotein ligand-1 binding. This compound is virtually non-fluorescent until cleavage by sulfatase enzymes, then emits blue fluorescence (450 nm) with excitation at 360 nm. Use of this compound in high-throughput fluorescence based screening led to the discovery of several potent beta-arylsulfotransferase IV (beta-AST-IV) inhibitors.
Solubility
Soluble in water at 5mg/ml
Notes
Excitation wavelength, 334 nm; emission wavelength, 370nm (pH 10.4). Reaction product: excitiation wavelength, 360nm; emission wavelength, 449nm
Chemical Identifiers
15220-11-8 | |
294.318 | |
CSOCSPXOODWGLJ-UHFFFAOYSA-M | |
5044226 | |
CC1=CC(=O)OC2=C1C=CC(=C2)OS(=O)(=O)[O-].[K+] |
C10H7KO6S | |
MFCD00016970 | |
4-methylumbelliferyl sulfate potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt, 4-methylumbelliferyl sulfate, potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt 1:1, potassium 4-methyl-2-oxo-2h-chromen-7-yl sulfate, potassium 4-methyl-2-oxochromen-7-yl sulfate, c10h7o6s.k, potassium 4-methyl-2-oxo-2h-1-benzopyran-7-yl sulphate, potassium 4-methylumbelliferyl sulfate | |
potassium;(4-methyl-2-oxochromen-7-yl) sulfate |
Specifications
15220-11-8 | |
C10H7KO6S | |
3803203 | |
Soluble in water at 5mg/ml | |
CC1=CC(=O)OC2=C1C=CC(=C2)OS(=O)(=O)[O-].[K+] | |
294.318 | |
294.32 | |
Powder |
White to Yellow | |
MFCD00016970 | |
4-methylumbelliferyl sulfate potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt, 4-methylumbelliferyl sulfate, potassium salt, 2h-1-benzopyran-2-one, 4-methyl-7-sulfooxy-, potassium salt 1:1, potassium 4-methyl-2-oxo-2h-chromen-7-yl sulfate, potassium 4-methyl-2-oxochromen-7-yl sulfate, c10h7o6s.k, potassium 4-methyl-2-oxo-2h-1-benzopyran-7-yl sulphate, potassium 4-methylumbelliferyl sulfate | |
CSOCSPXOODWGLJ-UHFFFAOYSA-M | |
potassium;(4-methyl-2-oxochromen-7-yl) sulfate | |
5044226 | |
98% | |
4-Methylumbelliferyl sulfate potassium salt |
Safety and Handling
missing translation for 'einecsNumber' : 239-272-2
missing translation for 'tsca' : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.