missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11328336
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Reactant for Suzuki-Miyaura cross-couplings, NHC-Iron-catalyzed aerobic oxidative aromatic esterification of aldehydes, Palladium-catalyzed oxidative Heck-type reactions.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145240-28-4
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.038
MDL Number MFCD02093926
InChI Key UGZUUTHZEATQAM-UHFFFAOYSA-N
Synonym 4-butylphenyl boronic acid, 4-n-butylphenylboronic acid, 4-butylphenylboronicacid, 4-butylbenzeneboronic acid, 4-n-butylbenzeneboronic acid, 4-butylphenyl boranediol, boronic acid, 4-butylphenyl, pubchem18523, acmc-209cue, 4-butylphenylboranic acid
PubChem CID 4100860
IUPAC Name (4-butylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCC)(O)O

Specifications

Melting Point 103°C to 106°C
Quantity 1 g
Beilstein 6920034
Solubility Information Soluble in methanol.
Formula Weight 178.04
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'