missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylphenol, 98%, Thermo Scientific Chemicals

58.09 € - 572.85 €

Chemical Identifiers

CAS 1638-22-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00041750
InChI Key CYYZDBDROVLTJU-UHFFFAOYSA-N
Synonym 4-n-butylphenol, p-butylphenol, phenol, 4-butyl, p-hydroxybutylbenzene, phenol, p-butyl, 1-hydroxy-4-n-butylbenzene, p-n-butylphenol, 4-butyl-phenol, 4-n-ethylphenol
PubChem CID 15420
ChEBI CHEBI:34437
IUPAC Name 4-butylphenol
SMILES CCCCC1=CC=C(C=C1)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11445183
View Documents
Thermo Scientific Alfa Aesar
A12812.06
5 g
58.09 €
5g
Please to purchase this item. Need a web account? Register with us today!
11455183
View Documents
Thermo Scientific Alfa Aesar
A12812.14
25 g
180.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
11465183
View Documents
Thermo Scientific Alfa Aesar
A12812.22
100 g
572.85 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-n-Butylphenol is used as an Intermediates of liquid Crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Butylphenol is used as an Intermediates of liquid Crystals.

Solubility
Not miscible with water.

Notes
When not in use, tightly seal the container and store in a dry, cool place. Avoid excessive heat and light. Incompatible to oxidizing agents. Stable if stored under proper conditions. Keep container dry. Keep away from heat. Mechanical exhaust required.
Specifications

Chemical Identifiers

1638-22-8
150.221
CYYZDBDROVLTJU-UHFFFAOYSA-N
15420
4-butylphenol
C10H14O
MFCD00041750
4-n-butylphenol, p-butylphenol, phenol, 4-butyl, p-hydroxybutylbenzene, phenol, p-butyl, 1-hydroxy-4-n-butylbenzene, p-n-butylphenol, 4-butyl-phenol, 4-n-ethylphenol
CHEBI:34437
CCCCC1=CC=C(C=C1)O

Specifications

1638-22-8
0.98
C10H14O
MFCD00041750
2042120
Not miscible with water.
CCCCC1=CC=C(C=C1)O
150.221
CHEBI:34437
98%
∼22°C
245°C
1.517
UN3145
4-n-butylphenol, p-butylphenol, phenol, 4-butyl, p-hydroxybutylbenzene, phenol, p-butyl, 1-hydroxy-4-n-butylbenzene, p-n-butylphenol, 4-butyl-phenol, 4-n-ethylphenol
CYYZDBDROVLTJU-UHFFFAOYSA-N
4-butylphenol
15420
150.22
4-n-Butylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 216-672-5

missing translation for 'rtecsNumber' : SJ8922500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.