missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylphenylboronic acid, 95%, Thermo Scientific™

Product Code. 11935871
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 145240-28-4
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.04
InChI Key UGZUUTHZEATQAM-UHFFFAOYSA-N
Synonym 4-butylphenyl boronic acid, 4-n-butylphenylboronic acid, 4-butylphenylboronicacid, 4-butylbenzeneboronic acid, 4-n-butylbenzeneboronic acid, 4-butylphenyl boranediol, boronic acid, 4-butylphenyl, pubchem18523, acmc-209cue, 4-butylphenylboranic acid
PubChem CID 4100860
IUPAC Name (4-butylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCC)(O)O

Specifications

Melting Point 91.0°C to 97.0°C
Assay Percent Range 94% min. (HPLC)
Linear Formula CH3(CH2)3C6H4B(OH)2
Quantity 5 g
Formula Weight 178.04
Percent Purity 95%
Chemical Name or Material 4-n-Butylphenylboronic acid
compliance-icons
Product Identifier
  • 4-n-Butylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'