Learn More
4-n-Decylbenzoyl chloride, 98%, Thermo Scientific Chemicals
41.70 € - 666.00 €
Chemical Identifiers
CAS | 54256-43-8 |
---|---|
Molecular Formula | C17H25ClO |
Molecular Weight (g/mol) | 280.836 |
MDL Number | MFCD00041724 |
InChI Key | HEWLDHSFOQXCSI-UHFFFAOYSA-N |
PubChem CID | 104714 |
IUPAC Name | 4-decylbenzoyl chloride |
SMILES | CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433457
|
Thermo Scientific Alfa Aesar
A17234.03 |
1 g | N/A | ||||||
11443457
|
Thermo Scientific Alfa Aesar
A17234.06 |
5 g |
166.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453457
|
Thermo Scientific Alfa Aesar
A17234.14 |
25 g |
666.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Employed as a pharmaceutical and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a pharmaceutical and chemical intermediate.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
Chemical Identifiers
54256-43-8 | |
280.836 | |
HEWLDHSFOQXCSI-UHFFFAOYSA-N | |
4-decylbenzoyl chloride |
C17H25ClO | |
MFCD00041724 | |
104714 | |
CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl |
Specifications
54256-43-8 | |
>110°C (230°F) | |
MFCD00041724 | |
Moisture sensitive | |
Reacts with water. | |
CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl | |
280.836 | |
280.84 | |
4-n-Decylbenzoyl chloride |
171°C to 173°C (2 mmHg) | |
C17H25ClO | |
UN3265 | |
4-n-decylbenzoyl chloride, benzoyl chloride, 4-decyl, p-decylbenzoyl chloride, 4-decylbenzoylchloride, p-n-decylbenzoyl chloride, benzoyl chloride,4-decyl, 4-n-decyl benzoyl chloride, 4-decylbenzoic acid chloride | |
HEWLDHSFOQXCSI-UHFFFAOYSA-N | |
4-decylbenzoyl chloride | |
104714 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 259-046-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only