missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Heptylacetophenone, 97%, Thermo Scientific™

Product Code. p-7044976
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11302806

Brand: Thermo Scientific Alfa Aesar L08959.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37593-03-6
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00038342
InChI Key UQBRZOXCKKBKDU-UHFFFAOYSA-N
Synonym 4'-heptylacetophenone, p-heptylacetophenone, ethanone, 1-4-heptylphenyl, p-n-heptylacetophenone, 1-4-heptylphenyl ethanone, 1-4-heptylphenyl ethan-1-one, 4'-n-heptylacetophenone, 4-heptylacetophenone, 1-acetyl-4-heptylbenzene, acmc-209iu8
PubChem CID 98879
IUPAC Name 1-(4-heptylphenyl)ethanone
SMILES CCCCCCCC1=CC=C(C=C1)C(=O)C

Specifications

Density 0.94
Boiling Point 155°C to 157°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.509
Quantity 1 g
Beilstein 1866760
Formula Weight 218.34
Percent Purity 97%
Chemical Name or Material 4'-n-Heptylacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.