missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptylphenol, 98+%, Thermo Scientific Chemicals

Product Code. 11488376
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1987-50-4
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00041751
InChI Key KNDDEFBFJLKPFE-UHFFFAOYSA-N
Synonym 4-n-heptylphenol, phenol, 4-heptyl, p-heptylphenol, heptanyl-p-phenol, p-hydroxyheptylbenzene, phenol, heptyl derivs., p-n-heptylphenol, unii-i743z35dvb, phenol, heptenylated, p-heptyl phenol
PubChem CID 16143
ChEBI CHEBI:34438
IUPAC Name 4-heptylphenol
SMILES CCCCCCCC1=CC=C(C=C1)O

Specifications

Melting Point 25°C to 28°C
Boiling Point 156°C (9 mmHg)
Refractive Index 1.507
Quantity 5 g
UN Number UN2430
Formula Weight 192.3
Percent Purity ≥98%
Chemical Name or Material 4-n-Heptylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.