missing translation for 'onlineSavingsMsg'
Learn More

4-n-Nonylphenol, 98+%, Thermo Scientific Chemicals

Product Code. 11487477
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

Solubility
Sparingly Soluble (0.020 g/L) at 25°C.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-40-5
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
MDL Number MFCD00002396
InChI Key IGFHQQFPSIBGKE-UHFFFAOYSA-N
Synonym 4-n-nonylphenol, p-nonylphenol, phenol, 4-nonyl, para-nonylphenol, p-n-nonylphenol, phenol, nonyl, phenol, p-nonyl, nonylphenol mixed, phenol, nonyl derivs., 4-nonyl phenol
PubChem CID 1752
ChEBI CHEBI:34440
IUPAC Name 4-nonylphenol
SMILES CCCCCCCCCC1=CC=C(C=C1)O

Specifications

Melting Point 43°C to 46°C
Density 0.937
Boiling Point 127°C to 130°C (27 mmHg)
Flash Point 150°C (302°F)
Refractive Index 1.511
Quantity 1 g
UN Number UN2430
Beilstein 2047450
Solubility Information Sparingly Soluble (0.020g/L) at 25°C.
Formula Weight 220.36
Percent Purity ≥98%
Chemical Name or Material 4-n-Nonylphenol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-n-Nonylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.
  • H361-Suspected of damaging fertility or the unborn child.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 4-n-Nonylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.