missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11443953
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49763-65-7
Molecular Formula C12H15ClO
Molecular Weight (g/mol) 210.701
MDL Number MFCD00000700
InChI Key FBBRKYLXMNQFQU-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride
PubChem CID 170812
IUPAC Name 4-pentylbenzoyl chloride
SMILES CCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.034
Boiling Point 144°C to 145°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 25 g
UN Number UN3265
Beilstein 2207887
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 210.71
Percent Purity 98%
Chemical Name or Material 4-n-Pentylbenzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'