missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Pentylbenzoyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
Specifications
Specifications
Density | 1.034 |
Boiling Point | 144°C to 145°C (10 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.53 |
Quantity | 100 g |
UN Number | UN3265 |
Beilstein | 2207887 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 210.71 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'