missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentyloxybenzeneboronic acid, 96%, Thermo Scientific™

41.30 € - 150.00 €

Chemical Identifiers

CAS 146449-90-3
Molecular Formula C11H17BO3
Molecular Weight (g/mol) 208.06
MDL Number MFCD04039035
InChI Key WHFKSJLAGACWMF-UHFFFAOYSA-N
Synonym 4-pentyloxyphenylboronic acid, 4-pentyloxyphenyl boronic acid, 4-pentyloxy phenylboronic acid, 4-n-pentyloxy benzeneboronic acid, 4-pentoxyphenyl boronic acid, 4-pentyloxyphenyl boronicacid, boronic acid, 4-pentyloxy phenyl, 4-n-pentyloxyphenyl boronic acid, pubchem9551, acmc-209cx1
PubChem CID 4684888
IUPAC Name (4-pentoxyphenyl)boronic acid
SMILES CCCCCOC1=CC=C(C=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15469797
View Documents
Thermo Scientific Alfa Aesar
H53026.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15479797
View Documents
Thermo Scientific Alfa Aesar
H53026.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

146449-90-3
208.06
WHFKSJLAGACWMF-UHFFFAOYSA-N
4684888
CCCCCOC1=CC=C(C=C1)B(O)O
C11H17BO3
MFCD04039035
4-pentyloxyphenylboronic acid, 4-pentyloxyphenyl boronic acid, 4-pentyloxy phenylboronic acid, 4-n-pentyloxy benzeneboronic acid, 4-pentoxyphenyl boronic acid, 4-pentyloxyphenyl boronicacid, boronic acid, 4-pentyloxy phenyl, 4-n-pentyloxyphenyl boronic acid, pubchem9551, acmc-209cx1
(4-pentoxyphenyl)boronic acid

Specifications

146449-90-3
C11H17BO3
4-pentyloxyphenylboronic acid, 4-pentyloxyphenyl boronic acid, 4-pentyloxy phenylboronic acid, 4-n-pentyloxy benzeneboronic acid, 4-pentoxyphenyl boronic acid, 4-pentyloxyphenyl boronicacid, boronic acid, 4-pentyloxy phenyl, 4-n-pentyloxyphenyl boronic acid, pubchem9551, acmc-209cx1
CCCCCOC1=CC=C(C=C1)B(O)O
208.06
208.06
4-n-Pentyloxybenzeneboronic acid
98°C to 100°C
MFCD04039035
WHFKSJLAGACWMF-UHFFFAOYSA-N
(4-pentoxyphenyl)boronic acid
4684888
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications