Learn More
4-n-Propylpiperidine, 95%, Thermo Scientific Chemicals
57.99 € - 144.00 €
Chemical Identifiers
CAS | 22398-09-0 |
---|---|
Molecular Formula | C8H17N |
Molecular Weight (g/mol) | 127.231 |
MDL Number | MFCD00023151 |
InChI Key | RQGBFVLTFYRYKB-UHFFFAOYSA-N |
Synonym | 4-n-propylpiperidine, piperidine, 4-propyl, 4-propyl-piperidine, pubchem2452, 4-prop-1-yl piperidine, 5-20-04-00208 beilstein handbook reference |
PubChem CID | 31169 |
IUPAC Name | 4-propylpiperidine |
SMILES | CCCC1CCNCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15453317
|
Thermo Scientific Alfa Aesar
H25848.MD |
250 mg |
57.99 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495580
|
Thermo Scientific Alfa Aesar
H25848.03 |
1 g |
144.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22398-09-0 | |
127.231 | |
RQGBFVLTFYRYKB-UHFFFAOYSA-N | |
31169 | |
CCCC1CCNCC1 |
C8H17N | |
MFCD00023151 | |
4-n-propylpiperidine, piperidine, 4-propyl, 4-propyl-piperidine, pubchem2452, 4-prop-1-yl piperidine, 5-20-04-00208 beilstein handbook reference | |
4-propylpiperidine |
Specifications
22398-09-0 | |
178°C to 180°C | |
Amine-like | |
1.453 | |
UN2733 | |
4-n-propylpiperidine, piperidine, 4-propyl, 4-propyl-piperidine, pubchem2452, 4-prop-1-yl piperidine, 5-20-04-00208 beilstein handbook reference | |
CCCC1CCNCC1 | |
127.231 | |
127.23 | |
4-n-Propylpiperidine |
0.864 | |
48°C (118°F) | |
C8H17N | |
MFCD00023151 | |
Air Sensitive | |
RQGBFVLTFYRYKB-UHFFFAOYSA-N | |
4-propylpiperidine | |
31169 | |
95% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 244-956-9
missing translation for 'rtecsNumber' : TN3740005
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only