missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitro-1H-pyrazole-3-carboxylic acid, 98%, Thermo Scientific Chemicals
21.80 € - 130.00 €
Chemical Identifiers
CAS | 5334-40-7 |
---|---|
Molecular Formula | C4H3N3O4 |
Molecular Weight (g/mol) | 157.085 |
MDL Number | MFCD00090899 |
InChI Key | ZMAXXOYJWZZQBK-UHFFFAOYSA-N |
Synonym | 4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic |
PubChem CID | 219739 |
SMILES | C1=NNC(=C1[N+](=O)[O-])C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356659
|
Thermo Scientific Alfa Aesar
H26224.03 |
1 g |
21.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366659
|
Thermo Scientific Alfa Aesar
H26224.09 |
10 g |
130.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5334-40-7 | |
157.085 | |
ZMAXXOYJWZZQBK-UHFFFAOYSA-N | |
219739 |
C4H3N3O4 | |
MFCD00090899 | |
4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic | |
C1=NNC(=C1[N+](=O)[O-])C(=O)O |
Specifications
5334-40-7 | |
C4H3N3O4 | |
4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic | |
C1=NNC(=C1[N+](=O)[O-])C(=O)O | |
219739 | |
98% |
∼195°C (decomposition) | |
MFCD00090899 | |
ZMAXXOYJWZZQBK-UHFFFAOYSA-N | |
157.085 | |
157.08 | |
4-Nitro-1H-pyrazole-3-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only