missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetanilide, 98%, Thermo Scientific Chemicals

Product Code. 11410014
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-04-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00007303
InChI Key NQRLPDFELNCFHW-UHFFFAOYSA-N
Synonym 4'-nitroacetanilide, n-4-nitrophenyl acetamide, 4-nitroacetanilide, p-nitroacetanilide, acetamide, n-4-nitrophenyl, n-acetyl-4-nitroaniline, p-acetamidonitrobenzene, acetanilide, p-nitro, p-nitrophenylacetanilide, n-acetyl-p-nitroaniline
PubChem CID 7691
IUPAC Name N-(4-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 215°C to 217°C
Quantity 5 g
Beilstein 2211962
Merck Index 14,6581
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 4'-Nitroacetanilide
compliance-icons
Product Identifier
  • 4'-Nitroacetanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4'-Nitroacetanilide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.