missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetanilide, 98%, Thermo Scientific Chemicals

Product Code. 11420014
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-04-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00007303
InChI Key NQRLPDFELNCFHW-UHFFFAOYSA-N
Synonym 4'-nitroacetanilide, n-4-nitrophenyl acetamide, 4-nitroacetanilide, p-nitroacetanilide, acetamide, n-4-nitrophenyl, n-acetyl-4-nitroaniline, p-acetamidonitrobenzene, acetanilide, p-nitro, p-nitrophenylacetanilide, n-acetyl-p-nitroaniline
PubChem CID 7691
IUPAC Name N-(4-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 215°C to 217°C
Quantity 25 g
Beilstein 2211962
Merck Index 14,6581
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 4'-Nitroacetanilide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'