Learn More
4-Nitroaniline, 98%, Thermo Scientific Chemicals
41.20 € - 330.00 €
Chemical Identifiers
CAS | 100-01-6 |
---|---|
Molecular Formula | C6H6N2O2 |
Molecular Weight (g/mol) | 138.13 |
MDL Number | MFCD00007858 |
InChI Key | TYMLOMAKGOJONV-UHFFFAOYSA-N |
Synonym | p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p |
PubChem CID | 7475 |
ChEBI | CHEBI:17064 |
IUPAC Name | 4-nitroaniline |
SMILES | NC1=CC=C(C=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11414183
|
Thermo Scientific Alfa Aesar
A10369.30 |
250 g |
41.20 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404183
|
Thermo Scientific Alfa Aesar
A10369.0B |
1000 g |
105.00 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424183
|
Thermo Scientific Alfa Aesar
A10369.0I |
5000 g |
330.00 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.
Solubility
Soluble in hot water and alcohol
Notes
Store at 4°C. Incompatible with oxidizing agents.
Chemical Identifiers
100-01-6 | |
138.13 | |
TYMLOMAKGOJONV-UHFFFAOYSA-N | |
7475 | |
4-nitroaniline |
C6H6N2O2 | |
MFCD00007858 | |
p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p | |
CHEBI:17064 | |
NC1=CC=C(C=C1)[N+]([O-])=O |
Specifications
100-01-6 | |
1.437 | |
165°C (329°F) | |
C6H6N2O2 | |
UN1661 | |
14,6584 | |
Soluble in hot water and alcohol | |
NC1=CC=C(C=C1)[N+]([O-])=O | |
138.13 | |
CHEBI:17064 | |
98% |
147°C to 149°C | |
260°C (100 mmHg) | |
Amine-like | |
MFCD00007858 | |
508690 | |
p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p | |
TYMLOMAKGOJONV-UHFFFAOYSA-N | |
4-nitroaniline | |
7475 | |
138.13 | |
4-Nitroaniline |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES
missing translation for 'einecsNumber' : 202-810-1
missing translation for 'rtecsNumber' : BY7000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only