missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.
Solubility
Soluble in water, ethanol, benzene, glacial acetic acid. Slightly soluble in ether.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.
Solubility
Soluble in water, ethanol, benzene, glacial acetic acid. Slightly soluble in ether.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
Specifications
Melting Point | 104°C to 106°C |
Quantity | 25 g |
Beilstein | 386796 |
Sensitivity | Air Sensitive |
Merck Index | 14,6587 |
Solubility Information | Soluble in water,ethanol,benzene,glacial acetic acid. Slightly soluble in ether. |
Formula Weight | 151.12 |
Percent Purity | 99% |
Chemical Name or Material | 4-Nitrobenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'