missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenediazonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

33.17 € - 237.18 €

Chemical Identifiers

CAS 456-27-9
Molecular Formula C6H4BF4N3O2
Molecular Weight (g/mol) 236.92
MDL Number MFCD00012005
InChI Key AFULQCYCQAHYIP-UHFFFAOYSA-N
Synonym 4-nitrobenzenediazonium tetrafluoroborate, ccris 3641, unii-2uxo615r4o, p-nitrobenzenediazonium tetrafluoroborate, benzenediazonium, 4-nitro-, tetrafluoroborate 1-, p-nitrophenyldiazonium fluoborate, p-nitrobenzenediazonium fluoborate, 4-nitrobenzenediazonium fluoborate, p-nitrobenzenediazonium fluoroborate, 4-nitrobenzenediazonium fluoroborateShow MoreShow Less
PubChem CID 68006
IUPAC Name 4-nitrobenzenediazonium;tetrafluoroborate
SMILES F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431538
SDS View Documents
Thermo Scientific Alfa Aesar
H55315.03
1 g
33.17 €
1g
Please to purchase this item. Need a web account? Register with us today!
15441538
SDS View Documents
Thermo Scientific Alfa Aesar
H55315.06
5 g
69.75 €
5g
Please to purchase this item. Need a web account? Register with us today!
15451538
SDS View Documents
Thermo Scientific Alfa Aesar
H55315.14
25 g
237.18 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4'-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4′-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

Solubility
Soluble in acetonitrile and dimethyl sulfoxide.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 207-261-1

missing translation for 'rtecsNumber' : CZ1723000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrobenzenediazonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.