missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

24.62 € - 157.79 €

Chemical Identifiers

CAS 122-04-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007345
InChI Key SKDHHIUENRGTHK-UHFFFAOYSA-N
Synonym p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
PubChem CID 8502
IUPAC Name 4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374689
View Documents
Thermo Scientific Alfa Aesar
A12543.09
10 g
24.62 €
10g
Please to purchase this item. Need a web account? Register with us today!
11484224
View Documents
Thermo Scientific Alfa Aesar
A12543.30
250 g
47.34 €
250g
Please to purchase this item. Need a web account? Register with us today!
11474224
View Documents
Thermo Scientific Alfa Aesar
A12543.0B
1000 g
157.79 €
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

Solubility
Soluble in terahydrofuran, dichloromethane, chloroform and pyridine.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
Specifications

Chemical Identifiers

122-04-3
185.56
SKDHHIUENRGTHK-UHFFFAOYSA-N
8502
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
C7H4ClNO3
MFCD00007345
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
4-nitrobenzoyl chloride

Specifications

122-04-3
1.53
102°C (215°F)
C7H4ClNO3
UN3261
Moisture sensitive
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
SKDHHIUENRGTHK-UHFFFAOYSA-N
4-nitrobenzoyl chloride
8502
98%
71°C to 74°C
155°C (15 mmHg)
Characteristic
MFCD00007345
473192
14,6590
Soluble in terahydrofuran,dichloromethane,chloroform and pyridine.
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
185.56
185.57
4-Nitrobenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 204-517-4

missing translation for 'rtecsNumber' : DM6651000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.