missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 10497920

4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10497920
100 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10497920

Brand: Thermo Scientific Acros 150431000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-04-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007345
InChI Key SKDHHIUENRGTHK-UHFFFAOYSA-N
Synonym p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
PubChem CID 8502
IUPAC Name 4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 74°C
Color Yellow
Boiling Point 202°C to 205°C (105.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4COCl
Quantity 100 g
Beilstein 09, 394
Fieser 01,736
Merck Index 15, 6677
Solubility Information Solubility in water: decomposes in water. Other solubilities: decomposes in alcohol,soluble in diethyl ether
Formula Weight 185.57
Percent Purity 98%
Physical Form Crystals or Flakes
Chemical Name or Material 4-Nitrobenzoyl chloride, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466852 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number150435000,150430050,150432500,150431000
Lot NumberA0444659 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number150435000,150430050,150432500,150431000
Lot NumberA0400437 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150435000,150430050,150432500,150431000
Lot NumberA0394231 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150435000,150430050,150432500,150431000
Lot NumberA0387601 Certificate TypeCertificate of Analysis Date14/05/2022 Catalog Number150435000,150430050,150432500,150431000
compliance-icons
Product Identifier
  • 4-Nitrobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.