missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroguaiacol, 97%, Thermo Scientific Chemicals

Product Code. 10463015
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3251-56-7
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00012143
InChI Key IZLVFLOBTPURLP-UHFFFAOYSA-N
Synonym 4-nitroguaiacol, phenol, 2-methoxy-4-nitro, guaiacol, 4-nitro, 2-hydroxy-5-nitroanisole, 3-nitro-6-hydroxyanisole, 4-hydroxy-3-methoxynitrobenzene, phenol, o-methoxy-p-nitro, o-methoxy-p-nitrophenol, 2-methoxy-4-nitro-phenol, 4-nitrogualacol
PubChem CID 76738
ChEBI CHEBI:81050
IUPAC Name 2-methoxy-4-nitrophenol
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 99.0°C to 104.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula O2NC6H3(OCH3)OH
Quantity 25 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 169.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Nitroguaiacol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'