missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenethyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 15505056
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-27-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
InChI Key IKMXRUOZUUKSON-UHFFFAOYSA-N
Synonym 4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
PubChem CID 7494
IUPAC Name 2-(4-nitrophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)[N+](=O)[O-]

Specifications

Melting Point 56.0°C to 57.0°C
Boiling Point 148.0°C to 149.0°C (2.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Linear Formula O2NC6H4CH2CH2OH
Quantity 10 g
Beilstein 06,II,238
Formula Weight 167.16
Percent Purity 99%
Chemical Name or Material 4-Nitrophenethyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'