missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Melting Point | >112.0°C |
Color | Yellow to Brown |
Boiling Point | 279.0°C |
Flash Point | 169°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Plastic drum |
Linear Formula | O2NC6H4OH |
Quantity | 5 kg |
Beilstein | 06, 226 |
Show More |
Safety and Handling
Product Identifier
- 4-Nitrophenol
- Danger
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H312-Harmful in contact with skin.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction