missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl acetate, 97%

Product Code. 10113660 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10113660

missing translation for 'mfr': Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-03-5
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007326
InChI Key QAUUDNIGJSLPSX-UHFFFAOYSA-N
Synonym p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester
PubChem CID 13243
ChEBI CHEBI:82635
SMILES CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 76.0°C to 79.0°C
Color Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3CO2C6H4NO2
Quantity 25 g
Beilstein 06, 233
Fieser 02,297
Solubility Information Solubility in water: insoluble
IUPAC Name (4-nitrophenyl) acetate
Formula Weight 181.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Nitrophenyl acetate, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.