missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11357506
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

Solubility
It hydrolyzes with water. Soluble in toluene.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2131-61-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD00007307
InChI Key NXHSSIGRWJENBH-UHFFFAOYSA-N
Synonym 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate
PubChem CID 75044
IUPAC Name 1-isothiocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=S)[N+](=O)[O-]

Specifications

Melting Point 105°C to 110°C
Boiling Point 137°C to 138°C (11 mmHg)
Odor Irritating
Quantity 250 g
UN Number UN2923
Beilstein 640027
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water. Soluble in toluene.
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'