missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals Product Code.: 11378565

4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals

Product Code. 11378565
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11378565

Brand: Thermo Scientific Alfa Aesar L12022.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1956-10-1
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00024665
InChI Key GGIDEJQGAZSTES-UHFFFAOYSA-N

Specifications

Density 1.082
Refractive Index 1.512
Quantity 5 g
Synonym 4-nitrophenyl caprylate, 4-nitrophenyloctanoate, p-nitrophenyl caprylate, octanoic acid, 4-nitrophenyl ester, 4-nitrophenyl octanoate, caprylic acid 4-nitrophenyl ester, octanoic acid 4-nitrophenyl ester, para-nitrophenyl octanoate, acmc-1bqka
SMILES CCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
IUPAC Name (4-nitrophenyl) octanoate
PubChem CID 97416
Formula Weight 265.31
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl octanoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249030 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog NumberL12022.06,L12022.03
Lot Number10243578 Certificate TypeCertificate of Analysis Date23/09/2022 Catalog NumberL12022.06,L12022.03
Lot Number10214107 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12022.06,L12022.03
Lot Number10174082 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12022.06,L12022.03
Lot Number10135988 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL12022.06,L12022.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenyl octanoate, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.