missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals Product Code.: 11393056

4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals

Product Code. 11393056
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11393056

Brand: Thermo Scientific Alfa Aesar L10896.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

Solubility
Insoluble in water. Soluble in CHCl3.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1492-30-4
Molecular Formula C22H35NO4
Molecular Weight (g/mol) 377.525
MDL Number MFCD00047732
InChI Key LVZSQWIWCANHPF-UHFFFAOYSA-N
Synonym 4-nitrophenyl palmitate, p-nitrophenyl palmitate, hexadecanoic acid 4-nitrophenyl ester, 4-nitrophenyl hexadecanoate, hexadecanoic acid, 4-nitrophenyl ester, 4-nitrophenylpalmitate, paranitrophenyl palmitate, para-nitrophenyl palmitate, p-nitrophenyl hexadecanoate, 4-nitrophenyl palmitate #
PubChem CID 73891
ChEBI CHEBI:85645
IUPAC Name (4-nitrophenyl) hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 65°C to 66°C
Quantity 1 g
Beilstein 1891754
Solubility Information Insoluble in water. Soluble in CHCl3.
Formula Weight 377.53
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl palmitate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253289 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog NumberL10896.03,L10896.06
Lot Number10248335 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog NumberL10896.03,L10896.06
Lot Number10242668 Certificate TypeCertificate of Analysis Date09/08/2022 Catalog NumberL10896.03,L10896.06
Lot Number10231677 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL10896.03,L10896.06
Lot Number10207076 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL10896.03,L10896.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.