missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl phosphate disodium salt hexahydrate, 96%, Thermo Scientific Chemicals
48.40 € - 487.00 €
Chemical Identifiers
CAS | 333338-18-4 |
---|---|
Molecular Formula | C6H4NNa2O6P |
Molecular Weight (g/mol) | 263.05 |
MDL Number | MFCD00007319 |
InChI Key | VIYFPAMJCJLZKD-UHFFFAOYSA-L |
Synonym | pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system |
PubChem CID | 77949 |
IUPAC Name | disodium;(4-nitrophenyl) phosphate |
SMILES | [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11495053
|
Thermo Scientific Alfa Aesar
A12310.06 |
5 g |
48.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11405063
|
Thermo Scientific Alfa Aesar
A12310.14 |
25 g |
144.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415063
|
Thermo Scientific Alfa Aesar
A12310.22 |
100 g |
487.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSubstrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
Solubility
Miscible with water. Immiscible with ethanol and acetone.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Chemical Identifiers
333338-18-4 | |
263.05 | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
77949 | |
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 |
C6H4NNa2O6P | |
MFCD00007319 | |
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
disodium;(4-nitrophenyl) phosphate |
Specifications
333338-18-4 | |
C6H4NNa2O6P | |
5324661 | |
Miscible with water. Immiscible with ethanol and acetone. | |
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 | |
263.05 | |
371.15 (263.06 Anhydrous) | |
4-Nitrophenyl phosphate disodium salt hexahydrate |
>300°C | |
MFCD00007319 | |
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
disodium;(4-nitrophenyl) phosphate | |
77949 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 224-246-5
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only