missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals Product Code.: 15497829

4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals

Product Code. 15497829
1 g
Click to view available options
Quantity:
100 mg
500 mg
1 g
This item is not returnable. View return policy

Product Code. 15497829

Brand: Thermo Scientific Alfa Aesar J66465.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.

Solubility
Soluble in water

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21064-69-7
Molecular Formula C11H17N2O6P
Molecular Weight (g/mol) 304.24
MDL Number MFCD00077834
InChI Key NAIXASFEPQPICN-UHFFFAOYSA-N
Synonym p-nitrophenylphosphorylcholine, o-4-nitrophenylphosphoryl choline, 4-nitrophenylphosphorylcholine, 4-nitrophenyl 2-trimethylammonio ethyl phosphate, 4-nitrophenylphorylcholine, o-4-nitrophenyl-phosphoryl choline, 4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate, 2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner saltShow MoreShow Less
PubChem CID 4307994
IUPAC Name (4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate
SMILES C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 233°C to 235°C
Color Yellow
Quantity 1 g
Beilstein 1895422
Sensitivity Light and moisture sensitive
Solubility Information Soluble in water
Formula Weight 304.24
Physical Form Solid
Chemical Name or Material 4-Nitrophenylphosphorylcholine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN19L004 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberJ66465.ME,J66465.03,J66465.MC
Lot NumberW11K013 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberJ66465.ME,J66465.03,J66465.MC
Lot NumberN07K023 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog NumberJ66465.ME,J66465.03,J66465.MC
Lot NumberZ09J009 Certificate TypeCertificate of Analysis Date28/12/2023 Catalog NumberJ66465.ME,J66465.03,J66465.MC
Lot NumberY09J019 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberJ66465.ME,J66465.03,J66465.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.