missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene, 99%, Thermo Scientific Chemicals

Product Code. 10003260
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14735-70-7
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD20481984
InChI Key WBZQHVKGKQXWMW-UHFFFAOYNA-N
Synonym 3-aza-tricyclo 4.2.1.0 2,5 non-7-en-4-one
PubChem CID 130862428
IUPAC Name 3-azatricyclo[4.2.1.0²,⁵]non-7-en-4-one
SMILES O=C1NC2C1C1CC2C=C1

Specifications

Melting Point 117.0°C to 122.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in benzene and ethanol
Formula Weight 135.17
Percent Purity 99%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material 4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'